Please login to your online account to display your discounted pricing

chroman-8-sulfonyl chloride, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC76103CB

 View more versions of this product

Catalog No. CC76103CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 232.68
Quantity 250mg
Assay 97%
CAS 104897015
Chemical Name or Material chroman-8-sulfonyl chloride
Packaging Amber Glass Bottle
Molecular Formula C9H9ClO3S
Health Hazard 1 GHS Signal Word: Danger

Specifications

Chemical Name or Material chroman-8-sulfonyl chloride
Assay 97%
Molecular Formula C9H9ClO3S
Formula Weight 232.68
Accessories

GHS Signal Word: Danger

SDS & Safety Documents