Please login to your online account to display your discounted pricing

chroman-8-sulfonyl chloride, Maybridge

Chemical Identifiers

Chemical Name or Material chroman-8-sulfonyl chloride
Assay 97%
Molecular Formula C9H9ClO3S
Formula Weight 232.68
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC76103CB

  SDS
fisher scientific company
CC76103CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC76103DA

  SDS
fisher scientific company
CC76103DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC76103DE

  SDS
fisher scientific company
CC76103DE
Amber Glass Bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 232.68
Quantity 250mg
Assay 97%
CAS 104897015
Molecular Formula C9H9ClO3S
Chemical Name or Material chroman-8-sulfonyl chloride
Packaging Amber Glass Bottle
Health Hazard 1 GHS Signal Word: Danger
Safety and Handling

GHS Signal Word: Danger