Please login to your online account to display your discounted pricing

chroman-8-carbonyl chloride, Maybridge


Quantity 5g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC76102DE

 View more versions of this product

Catalog No. CC76102DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 196.63
Quantity 5g
Assay 90%
Molecular Formula C10H9ClO2
CAS 103456609
Chemical Name or Material chroman-8-carbonyl chloride
Packaging Amber Glass Bottle
Health Hazard 1 GHS Signal Word: Danger

Specifications

Chemical Name or Material chroman-8-carbonyl chloride
Assay 90%
Molecular Formula C10H9ClO2
Formula Weight 196.63
Accessories

GHS Signal Word: Danger

SDS & Safety Documents