Please login to your online account to display your discounted pricing

chroman-8-carbonyl chloride, Maybridge

Chemical Identifiers

Chemical Name or Material chroman-8-carbonyl chloride
Assay 90%
Molecular Formula C10H9ClO2
Formula Weight 196.63
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC76102DA

  SDS
fisher scientific company
CC76102DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC76102CB

  SDS
fisher scientific company
CC76102CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC76102DE

  SDS
fisher scientific company
CC76102DE
Amber Glass Bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 196.63
Quantity 1g
Assay 90%
Chemical Name or Material chroman-8-carbonyl chloride
Packaging Amber Glass Bottle
Molecular Formula C10H9ClO2
CAS 103456609
Health Hazard 1 GHS Signal Word: Danger
Safety and Handling

GHS Signal Word: Danger