missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chroman-6-ylmethylamine, 97%, Thermo Scientific™

Manufacturer:  Fisher Scientific Company CC32813DE

 View more versions of this product

Catalog No. CC32813DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Chroman-6-ylmethylamine
96.5
97%
MFCD08445722
1-3,4-dihydro-2h-1-benzopyran-6-yl methanamine, 1-3,4-dihydro-2h-chromen-6-yl methanamine, 3,4-dihydro-2h-1-benzopyran-6-ylmethanamine, 3,4-dihydro-2h-chromen-6-ylmethyl amine, c-chroman-6-yl-methylamine, c-chroman-6-ylmethylamine, chroman-6-ylmethanamine, chroman-6-ylmethylamine
IVHMTTRMDOTDKZ-UHFFFAOYSA-N
3,4-dihydro-2H-chromen-6-ylmethanamine
18524876
97%
55746-21-9
100.0
C10H13NO
Amber Glass Bottle
5g
C1CC2=C(C=CC(=C2)CN)OC1
163.22
163.22

Chemical Identifiers

55746-21-9
163.22
IVHMTTRMDOTDKZ-UHFFFAOYSA-N
18524876
C1CC2=C(C=CC(=C2)CN)OC1
C10H13NO
MFCD08445722
1-3,4-dihydro-2h-1-benzopyran-6-yl methanamine, 1-3,4-dihydro-2h-chromen-6-yl methanamine, 3,4-dihydro-2h-1-benzopyran-6-ylmethanamine, 3,4-dihydro-2h-chromen-6-ylmethyl amine, c-chroman-6-yl-methylamine, c-chroman-6-ylmethylamine, chroman-6-ylmethanamine, chroman-6-ylmethylamine
3,4-dihydro-2H-chromen-6-ylmethanamine
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents
Product Certifications
Promotions

Promotions