missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chroman-6-sulfonyl chloride, 97%, Maybridge

Manufacturer:  Fisher Scientific Company CC32803DA

 View more versions of this product

Catalog No. CC32803DA

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

232.69
97%
Chroman-6-sulfonyl chloride
96.5
95%
Amber Glass Bottle
1g
100.0
946409-11-6
MFCD09897729
C9H9ClO3S

Chemical Identifiers

946409-11-6
100.0
C9H9ClO3S
MFCD09897729
97%
96.5
95%
232.69
Chroman-6-sulfonyl chloride

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger