missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chroman-6-sulfonyl chloride, 97%, Maybridge

Chemical Identifiers

CAS 946409-11-6
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C9H9ClO3S
Formula Weight 232.69
MDL Number MFCD09897729
Chemical Name or Material Chroman-6-sulfonyl chloride
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC32803DA SDS Fisher Scientific Company
CC32803DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC32803CB SDS Fisher Scientific Company
CC32803CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

946409-11-6
100.0
C9H9ClO3S
MFCD09897729
97%
96.5
95%
232.69
Chroman-6-sulfonyl chloride

Specifications

232.69
100.0
96.5
95%
Amber Glass Bottle
97%
1g
946409-11-6
MFCD09897729
C9H9ClO3S
Chroman-6-sulfonyl chloride

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger