missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Chromacol Injection Caps for Storage Vials

Thermo Scientific™ Chromacol Injection Caps are offered in a range of thread sizes.

$41.31 - $41.31

Specifications

For Use With Storage Vials
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
10-800-151 Thermo Scientific™
20SC
20-400 Injection Cap Pack of 100 Each for $41.31
Specifications

Description

Thermo Scientific™ Chromacol Injection Caps are offered in a range of thread sizes.

Description & Specifications

Specifications

Storage Vials