Please login to your online account to display your discounted pricing

Tocris™ CHR 2797

Aminopeptidase inhibitor

$164.61 - $354.48

Specifications

Product Type CHR 2797
Inhibitors CHR 2797
Molecular Weight 406.48
Purity >98
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

359510R

 
tocris™
RDS359510
10mg Each for $354.48

35951R

 
tocris™
RDS35951
1mg Each for $164.61
Description & Specifications

Specifications

Product Type CHR 2797
Inhibitors CHR 2797
Molecular Weight 406.48
Purity >98

Aminopeptidase inhibitor (IC50 values are 100, 150, 220, > 1000, > 5000, > 10000 and > 30000nM for LAP, PuSA, aminopeptidase N, aminopeptidase B, PILSAP, LTA4 hydrolase and MetAP2 respectively). Potently inhibits tumor cell proliferation in a variety of tumor cell lines in vitro and in vivo.

  • Chemical Name: α-[[(2R)-2-[(1S)-1-Hydroxy-2-(hydroxyamino)-2-oxoethyl]-4-methyl-1-oxopentyl]amino]-benzeneacetic acid cyclopently ester
  • Formula: C21H30N2O6
  • CAS Number: 238750-77-1
  • Solubility: Soluble to 100mM in DMSO and to 50mM in ethanol
  • Purity: >98