Please login to your online account to display your discounted pricing

CHPG, Tocris


mGlu5 selective agonist

Quantity 10mg

Manufacturer: r & d systems  RDS104910

 View more versions of this product

Catalog No. 104910

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 201.61
Quantity 10mg
Chemical Name or Material CHPG
Assay Percent Range >99%
Molecular Formula C8H8NO3Cl
CAS 170846-74-9
Synonym (RS)-2-Chloro-5-hydroxyphenylglycine
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material CHPG
Assay Percent Range >99%
Molecular Formula C8H8NO3Cl
Formula Weight 201.61
Synonym (RS)-2-Chloro-5-hydroxyphenylglycine
Accessories

Store at Room Temperature