Please login to your online account to display your discounted pricing

CHPG, Tocris

mGlu5 selective agonist

$139.08 - $585.69

Chemical Identifiers

Chemical Name or Material CHPG
Assay Percent Range >99%
Molecular Formula C8H8NO3Cl
Formula Weight 201.61
Synonym (RS)-2-Chloro-5-hydroxyphenylglycine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

104910

 
r & d systems
RDS104910
10mg Each for $139.08

104950

 
r & d systems
RDS104950
50mg Each for $585.69
Description & Specifications

Specifications

Chemical Name or Material CHPG
CAS 170846-74-9
Recommended Storage Store at Room Temperature
Assay Percent Range >99%
Molecular Formula C8H8NO3Cl
Formula Weight 201.61
Synonym (RS)-2-Chloro-5-hydroxyphenylglycine
Quantity 10mg
Safety and Handling

Store at Room Temperature