missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tocris™ CHPG Sodium salt

Selective mGlu5 agonist. Sodium salt of CHPG (Cat. No. 1049)

$155.12 - $646.57

Specifications

Product Type CHPG Sodium salt
Inhibitors CHPG Sodium salt
Molecular Weight 223.59
Purity >99
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
369510R Tocris™
RDS369510
10mg Each for $155.12
369550R Tocris™
RDS369550
50mg Each for $646.57
Specifications

Description

Sodium salt of the selective mGlu5 agonist CHPG (Cat. No. 1049).
  • Chemical Name: (RS)-2-Chloro-5-hydroxyphenylglycine sodium salt
  • Formula: C8H7ClNNaO3
  • CAS Number: 1303993-73-8
  • Solubility: Soluble to 50mM in water and to 50mM in DMSO
  • Purity: >99
Description & Specifications

Specifications

CHPG Sodium salt
CHPG Sodium salt
223.59
>99