Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Chorionic Gonadotropin, Human/hCG Ab-5, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Chorionic Gonadotropin, Human/hCG Ab-5, Mouse Monoclonal Antibody.

$244.82 - $758.16

Specifications

Antigen Chorionic Gonadotropin, Human/hCG Ab-5
Clone CG04 + CG05
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

MS1065P0

 
thermo scientific™ lab vision™
MS1065P0
Purified Ab with BSA and Azide 100μL Each for $244.82

MS1065P1

 
thermo scientific™ lab vision™
MS1065P1
Purified Ab with BSA and Azide 500μL Each for $449.06

MS1065P1ABX

 
thermo scientific™ lab vision™
MS1065P1ABX
Ab without BSA/Azide (Frozen) 100μL Each for $578.43

MS1065PABX

 
thermo scientific™ lab vision™
MS1065PABX
Ab without BSA/Azide (Frozen) 200μL Each for $758.16

MS1065R7

 
thermo scientific™ lab vision™
MS1065R7
Ready-to-use for IHC (UVLP) 7mL Each for $355.31
Description & Specifications

Specifications

Antigen Chorionic Gonadotropin, Human/hCG Ab-5
Clone CG04 + CG05
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Host Species Murine
Regulatory Status RUO
Research Discipline Steroids Hormones and Growth Factors
Type Primary
Immunogen Purified Beta-subunit of HCG
Isotype IgG1 kappa

Ensure accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Chorionic Gonadotropin, Human/hCG Ab-5, Mouse Monoclonal Antibody.

Human Chorionic Gonadotropin (HCG) is a glycoprotein that is secreted in large quantities by normal trophoblasts. It is present only in trace amounts in non-pregnant urine and sera but rises sharply during pregnancy. HCG is composed of two non-identical, non-covalently linked polypeptide chains designated as the α- and Β-subunits. The α-subunit of HCG is nearly identical to that of thyroid stimulating hormone (TSH), follicle stimulating hormone (FSH), and luteinizing hormone (LH). A germ cell tumor which is positive for cytokeratin, placental alkaline phosphatase (PLAP), and HCG but negative for EMA and AFP is probably a choriocarcinoma.

Host Species: Mouse

Clone: CG04 + CG05

Isotype: IgG2a/κ + IgG1/κ

Species Reactivity: Human. Others not known

Immunogen: Purified β-subunit of hCG

Molecular Weight: 22 kDa (β-hCG)

Positive Control: Placenta or chorioca

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not optimum)

Note:

USA: RUO; Int'l: RUO