Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Chorionic Gonadotropin, Human/hCG Ab-5, Mouse Monoclonal Antibody


Ensure accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Chorionic Gonadotropin, Human/hCG Ab-5, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS1065P1ABX

 View more versions of this product

Catalog No. MS1065P1ABX

  • / Each
 

Description & Specifications

Specifications

Antigen Chorionic Gonadotropin, Human/hCG Ab-5
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CG04 + CG05
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Formulation Ab without BSA/Azide (Frozen)
Host Species Murine
Immunogen Purified Beta-subunit of HCG
Isotype IgG1 kappa
Quantity 100μL
Regulatory Status RUO
Research Discipline Steroids Hormones and Growth Factors
Type Primary
Monoclonal or Polyclonal Monoclonal
Target Species Human

Ensure accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Chorionic Gonadotropin, Human/hCG Ab-5, Mouse Monoclonal Antibody.

Human Chorionic Gonadotropin (HCG) is a glycoprotein that is secreted in large quantities by normal trophoblasts. It is present only in trace amounts in non-pregnant urine and sera but rises sharply during pregnancy. HCG is composed of two non-identical, non-covalently linked polypeptide chains designated as the α- and Β-subunits. The α-subunit of HCG is nearly identical to that of thyroid stimulating hormone (TSH), follicle stimulating hormone (FSH), and luteinizing hormone (LH). A germ cell tumor which is positive for cytokeratin, placental alkaline phosphatase (PLAP), and HCG but negative for EMA and AFP is probably a choriocarcinoma.

Host Species: Mouse

Clone: CG04 + CG05

Isotype: IgG2a/κ + IgG1/κ

Species Reactivity: Human. Others not known

Immunogen: Purified β-subunit of hCG

Molecular Weight: 22 kDa (β-hCG)

Positive Control: Placenta or chorioca

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not optimum)

Note:

USA: RUO; Int'l: RUO