Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Chorionic Gonadotropin, Human/hCG Ab-2, Rabbit Polyclonal Antibody

Ensure accurate and reproducible results in immunohistochemistry experiments with Thermo Scientific™ Chorionic Gonadotropin, Human/hCG Ab-2, Rabbit Polyclonal Antibody.

$206.08 - $699.84

Specifications

Antigen Chorionic Gonadotropin, Human/hCG Ab-2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Human
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

RB059A

 
thermo scientific™ lab vision™
RB059A
1mL Each for $699.84

RB059A0

 
thermo scientific
RB059A0
100μL Each for $206.08

RB059A1

 
thermo scientific™ lab vision™
RB059A1
500μL Each for $405.18

RB059R7

 
thermo scientific™ lab vision™
RB059R7
7mL Each for $344.92
Description & Specifications

Specifications

Antigen Chorionic Gonadotropin, Human/hCG Ab-2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Human
Host Species Rabbit
Regulatory Status RUO
Research Discipline Steroids Hormones and Growth Factors
Type Primary
Immunogen Purified Beta-subunit of HCG
Isotype IgG

Ensure accurate and reproducible results in immunohistochemistry experiments with Thermo Scientific™ Chorionic Gonadotropin, Human/hCG Ab-2, Rabbit Polyclonal Antibody.

Human Chorionic Gonadotropin (HCG) is a glycoprotein that is secreted in large quantities by normal trophoblasts. It is present only in trace amounts in non-pregnant urine and sera but rises sharply during pregnancy. HCG is composed of two non-identical, non-covalently linked polypeptide chains designated as the alpha- and Beta-subunits. The alpha-subunit of HCG is nearly identical to that of thyroid stimulating hormone (TSH), follicle stimulating hormone (FSH), and luteinizing hormone (LH). A germ cell tumor which is positive for cytokeratin, placental alkaline phosphatase (PLAP), and HCG but negative for EMA and AFP is probably a choriocarcinoma.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not known.

Immunogen: Purified β-subunit of hCG

Molecular Weight: 22 kDa

Positive Control: Placenta or chorioca

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin-fixed)

Note:

USA: RUO; Int'l: RUO