Please login to your online account to display your discounted pricing

Alfa Aesar™ Choline p-toluenesulfonate, 97%

$333.00 - $1,193.35

Chemical Identifiers

Chemical Name or Material Choline p-toluenesulfonate
CAS 55357-38-5
Molecular Formula C5H14NO
Formula Weight 275.36
MDL Number MFCD00079051
Synonym (2-Hydroxyethyl)trimethylammonium p-toluenesulfonate; Choline tosylate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2192918

  SDS
alfa aesar™
B2192918
50g Each for $333.00

AAB2192930

  SDS
alfa aesar™
B2192930
250g Each for $1,193.35
Description & Specifications

Specifications

Formula Weight 275.36
Synonym (2-Hydroxyethyl)trimethylammonium p-toluenesulfonate; Choline tosylate
Molecular Formula C5H14NO
Quantity 50g
CAS 55357-38-5
Chemical Name or Material Choline p-toluenesulfonate
Health Hazard 1 Warning
Assay 97%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00079051
Melting Point 105° to 108°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.