Please login to your online account to display your discounted pricing

Alfa Aesar™ Choline iodide, 98%

$32.20 - $185.15

Chemical Identifiers

Chemical Name or Material Choline iodide
CAS 17773-10-3
Molecular Formula HOCH2CH2N(CH3)3I
Formula Weight 231.08g/mol
MDL Number MFCD00011900
Beilstein 3912691
Synonym (2-Hydroxyethyl)trimethylammonium iodide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0086009

 
alfa aesar™
L00860-09
10g Each for $32.20

AAL0086030

 
alfa aesar™
L00860-30
250g Each for $185.15
Description & Specifications

Specifications

Melting Point ≈270°C
Quantity 10g
Chemical Name or Material Choline iodide
CAS 17773-10-3
Assay 98%
Molecular Formula HOCH2CH2N(CH3)3I
Formula Weight 231.08g/mol
MDL Number MFCD00011900
Beilstein 3912691
Synonym (2-Hydroxyethyl)trimethylammonium iodide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number GA4100000
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.