Please login to your online account to display your discounted pricing

Choline Hydroxide, ACROS Organics™


Quantity 10L
Packaging Plastic drum

Manufacturer: thermo fisher scientific  AC386720100

 View more versions of this product

Catalog No. AC386720100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Granular Powder
Assay 44.0 to 48.0% (Total base)
Formula Weight 121.18
Quantity 10L
Color White
Purity Grade Pure
Density 1.023
Health Hazard 1 GHS Signal Word: Danger
Molecular Formula C5H15NO2
Chemical Name or Material Choline hydroxide, 45 wt.% aqueous solution
Trimethylamine 300ppm (as packaged) max.
Packaging Plastic drum
Stabilizer 1800ppm ethylenediamine 3000ppm hydroxylamine
CAS 123-41-1
MDL Number MFCD00002831

Specifications

Chemical Name or Material Choline hydroxide, 45 wt.% aqueous solution
CAS 123-41-1
Purity Grade Pure
Assay 44.0 to 48.0% (Total base)
Molecular Formula C5H15NO2
Formula Weight 121.18
MDL Number MFCD00002831
Accessories

GHS Signal Word: Danger

SDS & Safety Documents