Learn More
Choline hydroxide, 46% w/w aq. soln., Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B24443AE
Description
Choline hydroxide is used as a precursor molecule for the neurotransmitter acetylcholine, it is involved in several functions including muscle and memory muscle control. It was used as a basic catalyst for aldol condensation reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCholine hydroxide is used as a precursor molecule for the neurotransmitter acetylcholine, it is involved in several functions including muscle and memory muscle control. It was used as a basic catalyst for aldol condensation reactions.
Solubility
Fully miscible in water.
Notes
Store in a cool, dry place in tightly closed container. Store away from oxidizing agents, air, acids.
Specifications
Choline hydroxide | |
1.073 | |
1.427 | |
MFCD00002831 | |
UN3267 | |
choline hydroxide, gossypine, sincaline, bursine, luridine, sinkalin, sinkaline, fagine, vidine, 2-hydroxy-n,n,n-trimethylethanaminium hydroxide | |
KIZQNNOULOCVDM-UHFFFAOYSA-M | |
2-hydroxyethyl(trimethyl)azanium;hydroxide | |
31255 | |
46% w/w aq. soln. |
123-41-1 | |
C5H15NO2 | |
HOCH2CH2N(CH3)3OH | |
100 mL | |
Air Sensitive | |
Fully miscible in water. | |
[OH-].C[N+](C)(C)CCO | |
121.18 | |
121.18 |
Chemical Identifiers
123-41-1 | |
121.18 | |
KIZQNNOULOCVDM-UHFFFAOYSA-M | |
31255 |
C5H15NO2 | |
MFCD00002831 | |
choline hydroxide, gossypine, sincaline, bursine, luridine, sinkalin, sinkaline, fagine, vidine, 2-hydroxy-n,n,n-trimethylethanaminium hydroxide | |
[OH-].C[N+](C)(C)CCO |
Safety and Handling
GHS H Statement
H314
Causes severe skin burns and eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.
EINECSNumber : 204-625-1
RTECSNumber : GA4025500
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only