missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Choline Chloride, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 110290050

 View more versions of this product

Catalog No. AC110290050



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Choline chloride
298.0°C to 304.0°C
Authentic
Plastic drum
HOCH2CH2N(CH3)3Cl
5 kg
15, 2211
Solubility in water: soluble. Other solubilities: soluble in ethanol
[Cl-].C[N+](C)(C)CCO
139.62
CHEBI:133341
99%
67-48-1
White
98.5 to 101.5% (on dry substance) (Argentometry)
C5H14ClNO
MFCD00011721
04, II, 722
choline chloride, hepacholine, biocolina, biocoline, lipotril, paresan, luridin chloride, choline hydrochloride, cholinium chloride, bilineurin chloride
SGMZJAMFUVOLNK-UHFFFAOYSA-M
2-hydroxyethyl(trimethyl)azanium;chloride
6209
139.62
Adhering Crystals

Chemical Identifiers

67-48-1
139.62
SGMZJAMFUVOLNK-UHFFFAOYSA-M
6209
2-hydroxyethyl(trimethyl)azanium;chloride
C5H14ClNO
MFCD00011721
choline chloride, hepacholine, biocolina, biocoline, lipotril, paresan, luridin chloride, choline hydrochloride, cholinium chloride, bilineurin chloride
CHEBI:133341
[Cl-].C[N+](C)(C)CCO
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 200-655-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only