Learn More
Choline bitartrate, 98+%, Thermo Scientific Chemicals
$85.68 - $475.63
Chemical Identifiers
CAS | 87-67-2 |
---|---|
Molecular Formula | C9H19NO7 |
Molecular Weight (g/mol) | 253.251 |
MDL Number | MFCD00036332 |
InChI Key | QWJSAWXRUVVRLH-UHFFFAOYSA-M |
Synonym | choline bitartrate, cholini bitartras, choline 3-carboxy-2,3-dihydroxypropanoate, 2-hydroxyethyl trimethylammonium bitartrate, choline tartrate 1:1, 2-hydroxy-n,n,n-trimethylethanaminium 3-carboxy-2,3-dihydroxypropanoate, cholinibitatratis, choline hydrogen l-+-tartrate, ethanaminium, 2-hydroxy-n,n,n-trimethyl-, 2r,3r-2,3-dihydroxybutanedioate 1:1 |
PubChem CID | 10198924 |
IUPAC Name | 2-hydroxyethyl(trimethyl)azanium;2,3,4-trihydroxy-4-oxobutanoate |
SMILES | C[N+](C)(C)CCO.C(C(C(=O)[O-])O)(C(=O)O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ6465730
|
Thermo Scientific Chemicals
J6465730 |
250 g |
Each for $85.68
|
|
|||||
AAJ64657A1
|
Thermo Scientific Chemicals
J64657A1 |
1 kg |
Each for $216.51
|
|
|||||
AAJ64657A4
|
Thermo Scientific Chemicals
J64657A4 |
2.5 kg |
Each for $475.63
|
|
|||||
Description
Choline Bitartrate is used by nerve cells to manufacture a chemical called acetylcholine, which effectively allows cells to communicate with each other. Choline is also heavily associated with neurological function and can help improve movement, coordination, and myofibril contraction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCholine Bitartrate is used by nerve cells to manufacture a chemical called acetylcholine, which effectively allows cells to communicate with each other. Choline is also heavily associated with neurological function and can help improve movement, coordination, and myofibril contraction.
Solubility
Soluble in alcohol and slightly soluble in water.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
87-67-2 | |
253.251 | |
QWJSAWXRUVVRLH-UHFFFAOYSA-M | |
10198924 | |
C[N+](C)(C)CCO.C(C(C(=O)[O-])O)(C(=O)O)O |
C9H19NO7 | |
MFCD00036332 | |
choline bitartrate, cholini bitartras, choline 3-carboxy-2,3-dihydroxypropanoate, 2-hydroxyethyl trimethylammonium bitartrate, choline tartrate 1:1, 2-hydroxy-n,n,n-trimethylethanaminium 3-carboxy-2,3-dihydroxypropanoate, cholinibitatratis, choline hydrogen l-+-tartrate, ethanaminium, 2-hydroxy-n,n,n-trimethyl-, 2r,3r-2,3-dihydroxybutanedioate 1:1 | |
2-hydroxyethyl(trimethyl)azanium;2,3,4-trihydroxy-4-oxobutanoate |
Specifications
87-67-2 | |
White | |
MFCD00036332 | |
Hygroscopic | |
Soluble in alcohol and slightly soluble in water. | |
C[N+](C)(C)CCO.C(C(C(=O)[O-])O)(C(=O)O)O | |
253.251 | |
253.25 | |
Powder |
151°C to 153°C | |
C9H19NO7 | |
250 g | |
choline bitartrate, cholini bitartras, choline 3-carboxy-2,3-dihydroxypropanoate, 2-hydroxyethyl trimethylammonium bitartrate, choline tartrate 1:1, 2-hydroxy-n,n,n-trimethylethanaminium 3-carboxy-2,3-dihydroxypropanoate, cholinibitatratis, choline hydrogen l-+-tartrate, ethanaminium, 2-hydroxy-n,n,n-trimethyl-, 2r,3r-2,3-dihydroxybutanedioate 1:1 | |
QWJSAWXRUVVRLH-UHFFFAOYSA-M | |
2-hydroxyethyl(trimethyl)azanium;2,3,4-trihydroxy-4-oxobutanoate | |
10198924 | |
≥98% | |
Choline bitartrate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 201-763-4
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only