missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cholic acid, 98+%

Manufacturer:  Alfa Aesar™ A1125736

 View more versions of this product

Catalog No. AAA1125736



Description

Description

Applications
Cholic acid is used as a preservative for food. It is widely used as an intermediate in the manufacture of pharmaceuticals. It finds use in biological research. It helps in detergents to soften biological fats. It helps in preventing and melting gallstones.

Solubility
Soluble in alcohols, acetone, dimethylsulfoxide, methanol. Insoluble in water.

Notes
Stable. Incompatible with other materials such as strong oxidizing agents.
Specifications

Specifications

Cholic acid
≥98%
MFCD00003672
14,2203
CC(CCC(=O)O)C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C
408.579
CHEBI:16359
81-25-4
C24H40O5
2822009
BHQCQFFYRZLCQQ-OELDTZBJSA-N
(4R)-4-[(3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid
221493
408.58

Chemical Identifiers

81-25-4
408.579
BHQCQFFYRZLCQQ-OELDTZBJSA-N
CHEBI:16359
CC(CCC(=O)O)C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C
C24H40O5
MFCD00003672
221493
(4R)-4-[(3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid
Safety and Handling

Safety and Handling

RTECSNumber : FZ9350000

TSCA : TSCA

SDS Documents