Please login to your online account to display your discounted pricing

Alfa Aesar™ Cholesteryl stearate, 96%

$39.32 - $675.03

Chemical Identifiers

Chemical Name or Material Cholesteryl stearate
CAS 35602-69-8
Molecular Formula C45H80O2
Formula Weight 653.13
MDL Number MFCD00003639
Beilstein 2068492
Synonym Cholesteryl octadecanoate; Octadecanoic acid cholesteryl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1477106

  SDS
alfa aesar™
A1477106
5g Each for $39.32

AAA1477114

  SDS
alfa aesar™
A1477114
25g Each for $110.30

AAA1477130

  SDS
alfa aesar™
A1477130
250g Each for $675.03
Description & Specifications

Specifications

Formula Weight 653.13
Chemical Name or Material Cholesteryl stearate
Assay 96%
Molecular Formula C45H80O2
MDL Number MFCD00003639
CAS 35602-69-8
Beilstein 2068492
Synonym Cholesteryl octadecanoate; Octadecanoic acid cholesteryl ester
Melting Point 76° to 79°C
Quantity 5g
Optical Rotation −23° (c=5 in Chloroform)

Applications
Cholesteryl stearate, is used as an emollient, emulsion stabilizer. It is also used as a chemical widely in lipid research.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Incompatible to strong oxidizing agents, strong acids, strong bases, strong reducing agents.