missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cholesteryl stearate, 96%

Manufacturer:  Alfa Aesar™ A1477130

 View more versions of this product

Catalog No. AAA1477130



Description

Description

Applications
Cholesteryl stearate, is used as an emollient, emulsion stabilizer. It is also used as a chemical widely in lipid research.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Incompatible to strong oxidizing agents, strong acids, strong bases, strong reducing agents.
Specifications

Specifications

Cholesteryl stearate
76°C to 79°C
35602-69-8
MFCD00003639
cholesteryl stearate, cholesterol stearate, cholestryl stearate, cholesteryl octadecanoate, 18:0 cholesteryl ester, cholest-5-en-3-beta-yl stearate, unii-i1q82n9dyq, cholest-5-en-3beta-yl octadecanoate, 5-cholesten-3.beta.-ol stearate, i1q82n9dyq
CCCCCCCCCCCCCCCCCC(=O)OC1CCC2(C3CCC4(C(C3CC=C2C1)CCC4C(C)CCCC(C)C)C)C
653.133
CHEBI:82750
96%
−23° (c=5 in Chloroform)
250g
C45H80O2
2068492
XHRPOTDGOASDJS-XNTGVSEISA-N
[(3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl] octadecanoate
118246
653.13

Chemical Identifiers

35602-69-8
653.133
XHRPOTDGOASDJS-XNTGVSEISA-N
118246
[(3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl] octadecanoate
C45H80O2
MFCD00003639
cholesteryl stearate, cholesterol stearate, cholestryl stearate, cholesteryl octadecanoate, 18:0 cholesteryl ester, cholest-5-en-3-beta-yl stearate, unii-i1q82n9dyq, cholest-5-en-3beta-yl octadecanoate, 5-cholesten-3.beta.-ol stearate, i1q82n9dyq
CHEBI:82750
CCCCCCCCCCCCCCCCCC(=O)OC1CCC2(C3CCC4(C(C3CC=C2C1)CCC4C(C)CCCC(C)C)C)C
SDS Documents