Please login to your online account to display your discounted pricing

Alfa Aesar™ Cholesteryl oleate

$45.09 - $544.91

Chemical Identifiers

Chemical Name or Material Cholesteryl oleate
CAS 303-43-5
Molecular Formula C45H78O2
Formula Weight 651.12
MDL Number MFCD00003645
Beilstein 2343071
Synonym Oleic acid cholesteryl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1137806

 
alfa aesar™
A1137806
5g Each for $45.09

AAA1137814

 
alfa aesar™
A1137814
25g Each for $158.54

AAA1137822

 
alfa aesar™
A1137822
100g Each for $544.91
Description & Specifications

Specifications

Formula Weight 651.12
Chemical Name or Material Cholesteryl oleate
TSCA TSCA
Molecular Formula C45H78O2
MDL Number MFCD00003645
CAS 303-43-5
Beilstein 2343071
Synonym Oleic acid cholesteryl ester
Melting Point 44° to 47°C
Quantity 5g
Odor Odorless
Optical Rotation −24° (c=1 in chloroform)

Applications
Cholesteryl oleate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff. It is also used in chemical research.

Solubility
It is soluble in chloroform: 0.1 g/Ml.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Incompatible to strong oxidizing agents.