missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cholesteryl chloride, 98%

$79.27 - $891.35

Chemical Identifiers

CAS 910-31-6
Molecular Formula C27H45Cl
Molecular Weight (g/mol) 405.107
MDL Number MFCD00003632
InChI Key OTVRYZXVVMZHHW-DGPGEHMFSA-N
Synonym cholesterylchloride
PubChem CID 131632519
IUPAC Name (8S,9R,10R,13R,14S,17R)-3-chloro-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthrene
SMILES CC(C)CCCC(C)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)Cl)C)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1043314 View Documents Alfa Aesar™
A1043314
25g Each for $79.27
 
AAA1043336 View Documents Alfa Aesar™
A1043336
500g Each for $891.35
 
AAA1043322 View Documents Alfa Aesar™
A1043322
100g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

910-31-6
405.107
OTVRYZXVVMZHHW-DGPGEHMFSA-N
131632519
CC(C)CCCC(C)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)Cl)C)C
C27H45Cl
MFCD00003632
cholesterylchloride
(8S,9R,10R,13R,14S,17R)-3-chloro-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthrene

Specifications

−26° (c=1 in benzene)
25g
C27H45Cl
2703655
OTVRYZXVVMZHHW-DGPGEHMFSA-N
(8S,9R,10R,13R,14S,17R)-3-chloro-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthrene
131632519
98%
94°C to 96°C
910-31-6
MFCD00003632
cholesterylchloride
CC(C)CCCC(C)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)Cl)C)C
405.107
405.11
Cholesteryl chloride
Safety and Handling

Safety and Handling

TSCA : TSCA