Please login to your online account to display your discounted pricing

Alfa Aesar™ Cholesteryl benzoate

$101.14 - $101.14

Chemical Identifiers

Chemical Name or Material Cholesteryl benzoate
CAS 604-32-0
Molecular Formula C34H50O2
Formula Weight 490.77
MDL Number MFCD00003635
Beilstein 3225829
Merck Index 14,2201
Synonym Benzoic acid cholesteryl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0359014

 
alfa aesar™
L0359014
25g Each for $101.14
Description & Specifications

Specifications

Formula Weight 490.77
Chemical Name or Material Cholesteryl benzoate
TSCA TSCA
Molecular Formula C34H50O2
MDL Number MFCD00003635
Beilstein 3225829
Merck Index 14,2201
Synonym Benzoic acid cholesteryl ester
Quantity 25g
Optical Rotation −15° (c=5 in Chloroform)
CAS 604-32-0
Melting Point 147° to 151°C
Odor Odorless

Applications
Cholesteryl benzoate is used as an emulsifying agent. It can be used with cholesteryl nonanoate and cholesteryl oleyl carbonate in some thermochromic liquid crystals. is used in some hair colors, make-ups, and some other cosmetic preparations.

Solubility
Soluble in Chloroform and Dichloromethane. Insoluble in water.

Notes
Store at -20°C. Incompatible with oxidizing agents.