Please login to your online account to display your discounted pricing

Alfa Aesar™ Cholesteryl acetate, 97+%

$61.78 - $518.32

Chemical Identifiers

Chemical Name or Material Cholesteryl acetate
CAS 604-35-3
Molecular Formula C29H48O2
Formula Weight 428.7
MDL Number MFCD00003636
Beilstein 2064235
Merck Index 14,2201
Synonym Acetic acid cholesteryl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1505214

  SDS
alfa aesar™
A1505214
25g Each for $61.78

AAA1505222

  SDS
alfa aesar™
A1505222
100g Each for $121.74

AAA1505236

  SDS
alfa aesar™
A1505236
500g Each for $518.32
Description & Specifications

Specifications

Formula Weight 428.7
Quantity 25g
CAS 604-35-3
Merck Index 14,2201
Optical Rotation −43° (c=5 in Chloroform)
TSCA TSCA
Chemical Name or Material Cholesteryl acetate
MDL Number MFCD00003636
Beilstein 2064235
Molecular Formula C29H48O2
Melting Point 112° to 114°C
Synonym Acetic acid cholesteryl ester
Assay ≥97%

Applications
Cholesteryl acetate is used in cosmetics, wrist watches, thermometers, propane tank volume indicators, video displays, pharmaceuticals.

Solubility
Soluble in chloroform, ethanol or ethyl acetate.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.