missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cholesteryl acetate, 97+%

Manufacturer:  Alfa Aesar™ A1505236

 View more versions of this product

Catalog No. AAA1505236



Description

Description

Applications
Cholesteryl acetate is used in cosmetics, wrist watches, thermometers, propane tank volume indicators, video displays, pharmaceuticals.

Solubility
Soluble in chloroform, ethanol or ethyl acetate.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

Cholesteryl acetate
112°C to 114°C
604-35-3
MFCD00003636
14,2201
XUGISPSHIFXEHZ-GGOZWAHDSA-N
[(10R,13R)-10,13-dimethyl-17-(6-methylheptan-2-yl)-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl] acetate
12360660
≥97%
−43° (c=5 in Chloroform)
500g
C29H48O2
2064235
cholesteryl acetate
CC(C)CCCC(C)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)OC(=O)C)C)C
428.701
428.7

Chemical Identifiers

604-35-3
428.701
XUGISPSHIFXEHZ-GGOZWAHDSA-N
12360660
CC(C)CCCC(C)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)OC(=O)C)C)C
C29H48O2
MFCD00003636
cholesteryl acetate
[(10R,13R)-10,13-dimethyl-17-(6-methylheptan-2-yl)-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl] acetate
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents