Please login to your online account to display your discounted pricing

Cholesterol, ≥99%, MP Biomedicals™


Quantity 100g

Manufacturer: mp biomedicals  10138280

 View more versions of this product

Catalog No. ICN10138280

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 386.64
Physical Form Powder
Color White
Quantity 100g
EINECS Number 200-353-2
Density 1.067g/mL
Synonym 3β-Hydroxy-5-cholestene
MDL Number MFCD00003646
CAS 57-88-5
Chemical Name or Material Cholesterol
Molecular Formula C27H46O
Assay Percent Range ≥99%
Recommended Storage Store at -0°C.

Specifications

Chemical Name or Material Cholesterol
CAS 57-88-5
Physical Form Powder
Assay Percent Range ≥99%
Molecular Formula C27H46O
Formula Weight 386.64
MDL Number MFCD00003646
Synonym 3β-Hydroxy-5-cholestene
Accessories

Store at -0°C.