Please login to your online account to display your discounted pricing

Alfa Aesar™ Cholesterol, 95%

$46.18 - $535.68

Chemical Identifiers

Chemical Name or Material Cholesterol
CAS 57-88-5
Molecular Formula C27H46O
Formula Weight 386.66
MDL Number MFCD00003646
Beilstein 1915888
Merck Index 14,2201
Synonym 5-Cholesten-3beta-ol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1147018

 
alfa aesar
A1147018
50g Each for $46.18

AAA114700B

 
alfa aesar™
A114700B
1000g Each for $535.68
Description & Specifications

Specifications

Formula Weight 386.66
Quantity 50g
TSCA TSCA
Assay 95%
MDL Number MFCD00003646
Beilstein 1915888
Merck Index 14,2201
RTECS Number FZ8400000
Chemical Name or Material Cholesterol
Assay Percent Range 95%
Molecular Formula C27H46O
CAS 57-88-5

Solubility
Soluble in alcohols, ether, chloroform, petroleum ether, pyridine, isopropyl myristate, acetone, ethylacetate, benzene, fats, oils and dioxane.

Notes
Incompatible with strong oxidizing agents, alkali metals, ammonia and peroxide.