Please login to your online account to display your discounted pricing

Alfa Aesar™ Cholecystokinin Flanking Peptide, Non-sulfated

For cellular and molecular biology applications

Specifications

Product Type Cholecystokinin Flanking Peptide, non-sulfated
Molecular Weight 1074.05
Storage Requirements -30° to -10°C
Sequence Ser-Ala-Glu-Glu-Tyr-Glu-Tyr-Pro-Ser
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66259EXD

 
alfa aesar™
J66259EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66259MCR

 
alfa aesar™
J66259MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Cholecystokinin Flanking Peptide, non-sulfated
Molecular Weight 1074.05
Storage Requirements -30° to -10°C
Sequence Ser-Ala-Glu-Glu-Tyr-Glu-Tyr-Pro-Ser
Format Solid
Regulatory Status RUO
Purification Method HPLC