Please login to your online account to display your discounted pricing

Alfa Aesar™ Cholecystokinin, CCK (27-33)


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J66940EXD

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ66940EXD

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Quantity 0.5mg
Format Solid
Sequence Tyr-Met-Gly-Trp-Met-Asp-Phe-NH2
Regulatory Status RUO
Storage Requirements -30° to -10°C
Common Name CCK Heptapeptide
Product Type Cholecystokinin, CCK (27-33)
Molecular Weight 948.12
Purification Method HPLC