Please login to your online account to display your discounted pricing

Alfa Aesar™ Cholecystokinin (26-33), Non-sulfated

For cellular and molecular biology applications

Specifications

Product Type Cholecystokinin (26-33), non-sulfated
Molecular Weight 1063.20
Storage Requirements -30° to -10°C
Sequence Asp-Tyr-Met-Gly-Trp-Met-Asp-Phe-NH2
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66378EXD

 
alfa aesar™
J66378EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66378MCR

 
alfa aesar™
J66378MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Cholecystokinin (26-33), non-sulfated
Molecular Weight 1063.20
Storage Requirements -30° to -10°C
Sequence Asp-Tyr-Met-Gly-Trp-Met-Asp-Phe-NH2
Format Solid
Common Name CCK Octapeptide (26-33), non-sulfated
Regulatory Status RUO
Purification Method HPLC