missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CHM 1, Tocris™

Potent antitumor agent; inducer of apoptosis

$199.90 - $838.64

Chemical Identifiers

Assay Percent Range >99%
Molecular Formula C16H10FNO3
Formula Weight 283.25
Synonym 6-(2-Fluorophenyl)-1, 3-dioxolo[4, 5-g]quinolin-8(5H)-one
Chemical Name or Material CHM 1
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
386850 R & D Systems
RDS386850
50mg Each for $838.64
386810 R & D Systems
RDS386810
10mg Each for $199.90
Specifications Description & Specifications

Chemical Identifiers

>99%
283.25
CHM 1
C16H10FNO3
6-(2-Fluorophenyl)-1, 3-dioxolo[4, 5-g]quinolin-8(5H)-one

Specifications

50mg
>99%
283.25
CHM 1
154554-41-3
C16H10FNO3
6-(2-Fluorophenyl)-1, 3-dioxolo[4, 5-g]quinolin-8(5H)-one

Safety and Handling

Recommended Storage : Store at Room Temperature