Please login to your online account to display your discounted pricing

Alfa Aesar™ Chlortetracycline hydrochloride, 10mg/mL in methanol, sterile-filtered


Quantity 25mL

Manufacturer: alfa aesar™  J67461AC

 View more versions of this product

Catalog No. AAJ67461AC

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

CAS 64-72-2
MDL Number MFCD00082440
Synonym Aureomycin
Health Hazard 2 GHS H Statement
H225-H311-H331-H370
Highly flammable liquid and vapor.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Chemical Name or Material Chlortetracycline hydrochloride
UN Number UN1230
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: METHANOL
Physical Form Liquid
RTECS Number QI7800000
Quantity 25mL
Health Hazard 1 Danger
Storage Note 1 -30° to -10°C
Name Note 10mg/mL in methanol, sterile-filtered

Specifications

Chemical Name or Material Chlortetracycline hydrochloride
CAS 64-72-2
Name Note 10mg/mL in methanol, sterile-filtered
MDL Number MFCD00082440
UN Number UN1230
Synonym Aureomycin
Accessories

Danger

GHS P Statement
P210-P260-P303+P361+P353-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H311-H331-H370
Highly flammable liquid and vapor.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.

-30° to -10°C

SDS & Safety Documents