Please login to your online account to display your discounted pricing

Alfa Aesar™ Chlortetracycline hydrochloride, 10mg/mL in methanol, sterile-filtered

Chemical Identifiers

Chemical Name or Material Chlortetracycline hydrochloride
CAS 64-72-2
Name Note 10mg/mL in methanol, sterile-filtered
MDL Number MFCD00082440
UN Number UN1230
Synonym Aureomycin
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67461AC

 
alfa aesar™
J67461AC
25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67461AD

 
alfa aesar™
J67461AD
50mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Chlortetracycline hydrochloride
MDL Number MFCD00082440
CAS 64-72-2
UN Number UN1230
Synonym Aureomycin
Health Hazard 2 GHS H Statement
H225-H311-H331-H370
Highly flammable liquid and vapor.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Name Note 10mg/mL in methanol, sterile-filtered
Physical Form Liquid
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1 -30° to -10°C
Quantity 25mL
DOT Information Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: METHANOL
RTECS Number QI7800000
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H311-H331-H370
Highly flammable liquid and vapor.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.

-30° to -10°C