missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Chlortetracycline hydrochloride, 10mg/mL in methanol, sterile-filtered

Manufacturer:  Alfa Aesar™ J67461AD

 View more versions of this product

Catalog No. AAJ67461AD



Specifications

Specifications

Chlortetracycline hydrochloride
64-72-2
MFCD00082440
Aureomycin
CC1(C2CC3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C(C=CC(=C41)Cl)O)O)O)O)C(=O)N)N(C)C)O.Cl
515.34
Liquid
10mg/mL in methanol, sterile-filtered
C22H24Cl2N2O8
UN1230
QYAPHLRPFNSDNH-CIVPRPTRSA-N
(4S,4aR,5aS,6S,12aR)-7-chloro-4-(dimethylamino)-1,6,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
66577600
50mL

Chemical Identifiers

64-72-2
515.34
QYAPHLRPFNSDNH-CIVPRPTRSA-N
66577600
CC1(C2CC3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C(C=CC(=C41)Cl)O)O)O)O)C(=O)N)N(C)C)O.Cl
C22H24Cl2N2O8
MFCD00082440
Aureomycin
(4S,4aR,5aS,6S,12aR)-7-chloro-4-(dimethylamino)-1,6,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H225-H311-H331-H370
Highly flammable liquid and vapor.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P307+P311-P311-P312-P330-P363-P370+P378q-P501c

H225-H301+H311+H331-H335-H370

DOTInformation : Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: METHANOL

EINECSNumber : 200-591-7

RTECSNumber : QI7800000

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

Documents