missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chloroxazone, 98%, ACROS Organics™

$151.81 - $151.81

Chemical Identifiers

CAS 95-25-0
Molecular Formula C7H4ClNO2
Molecular Weight (g/mol) 169.57
MDL Number MFCD00005717
InChI Key TZFWDZFKRBELIQ-UHFFFAOYSA-N
Synonym 5-chloro-2-benzoxazolone, biomioran, chloroxazone, chlorzoxazon, chlorzoxazone, escoflex, myoflexin, myoflexine, paraflex, pathorysin
PubChem CID 2733
ChEBI CHEBI:3655
IUPAC Name 5-chloro-3H-1,3-benzoxazol-2-one
SMILES C1=CC2=C(C=C1Cl)NC(=O)O2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC228950250
SDS View Documents
Thermo Fisher Scientific
AC228950250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC228951000
SDS View Documents
Thermo Fisher Scientific
228951000
100g Glass bottle
Each for $151.81
 
Description

Description

Specifications

Chemical Identifiers

95-25-0
169.57
TZFWDZFKRBELIQ-UHFFFAOYSA-N
2733
5-chloro-3H-1,3-benzoxazol-2-one
C7H4ClNO2
MFCD00005717
5-chloro-2-benzoxazolone, biomioran, chloroxazone, chlorzoxazon, chlorzoxazone, escoflex, myoflexin, myoflexine, paraflex, pathorysin
CHEBI:3655
C1=CC2=C(C=C1Cl)NC(=O)O2

Specifications

Authentic
1% max. (K.F.)
189°C to 194°C
95-25-0
C7H4ClNO2
27, 179
5-chloro-2-benzoxazolone, biomioran, chloroxazone, chlorzoxazon, chlorzoxazone, escoflex, myoflexin, myoflexine, paraflex, pathorysin
TZFWDZFKRBELIQ-UHFFFAOYSA-N
5-chloro-3H-1,3-benzoxazol-2-one
2733
169.57
98%
Glass bottle
Brown-Yellow to White
25g
98%
MFCD00005717
15, 2201
Solubility in water: soluble in hot water
C1=CC2=C(C=C1Cl)NC(=O)O2
169.57
CHEBI:3655
Powder
Chloroxazone, 98%
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 202-403-9

RTECSNumber : DM5250000

TSCA : TSCA

SDS