missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Chlorotris(triphenylphosphine)rhodium(I), 97%

Manufacturer:  Alfa Aesar™ 1046806

 View more versions of this product

Catalog No. AA1046806



Specifications

Specifications

Chlorotris(triphenylphosphine)rhodium(I)
Red
5g
C54H45ClP3Rh-
MFCD00010016
0fv534bkev
QBERHIJABFXGRZ-UHFFFAOYSA-M
rhodium;triphenylphosphane;chloride
84599
Micro Crystals
Odorless
~250°C (decomposition)
14694-95-2
RhCl[(C6H5)3P]3
14,10047
Soluble in most solvents (e.g. benzene, ethanol, chloroform, dichloromethane) but with phosphine dissociation; Reacts with O2 in solution
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.[Cl-].[Rh]
925.231
925.23
97%

Chemical Identifiers

14694-95-2
925.231
QBERHIJABFXGRZ-UHFFFAOYSA-M
84599
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.[Cl-].[Rh]
C54H45ClP3Rh-
MFCD00010016
0fv534bkev
rhodium;triphenylphosphane;chloride
Safety and Handling

Safety and Handling

TSCA : TSCA

storageNote1 : 2°C to 8°C

Documents