Please login to your online account to display your discounted pricing

chlorotripyrrolidinophosphonium hexafluorophosphate, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Chlorotripyrrolidinophosphonium hexafluorophosphate
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C12H24ClN3P·F6P
Formula Weight 421.73
Synonym PyCloP
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC438640250

 
thermo fisher scientific
AC438640250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC438640050

 
thermo fisher scientific
AC438640050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 421.73
Quantity 25g
Color White to yellow
Melting Point 144° to 149°C
Synonym PyCloP
Assay Percent Range 98%
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Elemental Analysis 33.33 to 35.03% C, 9.71 to 10.21% N
Chemical Name or Material Chlorotripyrrolidinophosphonium hexafluorophosphate
CAS 133894-48-1
Molecular Formula C12H24ClN3P·F6P
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Coupling reagent

Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.