Please login to your online account to display your discounted pricing

chlorotripropylsilane, 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC313930250

 View more versions of this product

Catalog No. AC313930250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 192.81
Physical Form Liquid
Quantity 25g
Color Colorless
Density 0.882g/mL
Refractive Index 1.3990 to 1.4410 (20°C, 589nm)
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Store in a dry place. Store in a closed container.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Protect from moisture.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material chlorotripropylsilane
CAS 995-25-5
Assay Percent Range 98%
Molecular Formula C9H21ClSi
Synonym Tripropylchlorosilane
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Infrared Spectrum Authentic

Specifications

Chemical Name or Material chlorotripropylsilane
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H21ClSi
Formula Weight 192.81
Synonym Tripropylchlorosilane
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Store in a dry place. Store in a closed container.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Protect from moisture.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

SDS & Safety Documents