Learn More
Chlorotriphenylsilane, 96%
CAS: 76-86-8 | C18H15ClSi | 294.85 g/mol
Supplier: Thermo Scientific Chemicals A1367814
Description
Chlorotriphenylsilane is used as a silylating agent. It is also used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsChlorotriphenylsilane is used as a silylating agent. It is also used as a pharmaceutical intermediate.
Solubility
Reacts with water.
Notes
Keep container tightly sealed. Store under dry inert gas. It is sensitive to moisture. Incompatible with bases and oxidizing agents.
Specifications
| 188°C to 189°C (3mmHg) | |
| 91°C to 96°C | |
| 25 g | |
| MFCD00000496 | |
| 1820487 | |
| triphenylsilyl chloride, triphenylchlorosilane, silane, chlorotriphenyl, triphenylsilicon chloride, chloro triphenyl silane, triphenylsilylchloride, chloro-tri phenyl silane, benzene, 1,1',1-chlorosilylidyne tris, clsiph3, ph3sicl | |
| MNKYQPOFRKPUAE-UHFFFAOYSA-N | |
| chloro(triphenyl)silane | |
| 6458 | |
| 96% | |
| Moisture sensitive |
| Chlorotriphenylsilane | |
| 76-86-8 | |
| C18H15ClSi | |
| UN3261 | |
| >200°C (392°F) | |
| Reacts with water. | |
| Cl[Si](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 294.85 | |
| 294.86 | |
| Pungent |
Chemical Identifiers
| 76-86-8 | |
| 294.85 | |
| MNKYQPOFRKPUAE-UHFFFAOYSA-N | |
| 6458 | |
| Cl[Si](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
| C18H15ClSi | |
| MFCD00000496 | |
| triphenylsilyl chloride, triphenylchlorosilane, silane, chlorotriphenyl, triphenylsilicon chloride, chloro triphenyl silane, triphenylsilylchloride, chloro-tri phenyl silane, benzene, 1,1',1-chlorosilylidyne tris, clsiph3, ph3sicl | |
| chloro(triphenyl)silane |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 200-989-0
RTECSNumber : VV2720000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only