missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Chlorotriphenylmethane, 98%

Manufacturer:  Alfa Aesar™ A1179936

 View more versions of this product

Catalog No. AAA1179936



Description

Description

Applications
Chlorotriphenylmethane is used as laboratory chemical. It acts as a protecting reagent for thiols, alcohols and amines. It is used in organic synthesis for initiating trityl group protection and for preparing S-trityl-L-cysteine (STLC), a inhibitor of Eg5 prevents mitotic progression.

Solubility
Insoluble in water. Soluble in chloroform, hexane, benzene, ether, acetone and tetrahydrofuran.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

Chlorotriphenylmethane
110°C to 114°C
76-83-5
(C6H5)3CCl
UN3261
trityl chloride, triphenylmethyl chloride, triphenylchloromethane, chlorotriphenylmethane, chloromethanetriyl tribenzene, triphenylmethylchloride, methane, chlorotriphenyl, triphenyl chloromethane, chlorodiphenylmethyl benzene, benzene, 1,1',1-chloromethylidyne tris
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)Cl
278.779
278.78
230°C to 235°C (20mmHg)
500g
C19H15Cl
MFCD00000813
397363
JBWKIWSBJXDJDT-UHFFFAOYSA-N
[chloro(diphenyl)methyl]benzene
6456
98%

Chemical Identifiers

76-83-5
278.779
JBWKIWSBJXDJDT-UHFFFAOYSA-N
6456
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)Cl
C19H15Cl
MFCD00000813
trityl chloride, triphenylmethyl chloride, triphenylchloromethane, chlorotriphenylmethane, chloromethanetriyl tribenzene, triphenylmethylchloride, methane, chlorotriphenyl, triphenyl chloromethane, chlorodiphenylmethyl benzene, benzene, 1,1',1-chloromethylidyne tris
[chloro(diphenyl)methyl]benzene
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P280-P273-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

RTECSNumber : PA6450000

TSCA : TSCA

SDS Documents