missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

chlorotrimethylsilane, 1M Solution in Dichloromethane, AcroSeal™, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC381601000

 View more versions of this product

Catalog No. AC381601000


 

Specifications Description & Specifications

Specifications

108.64
15, 2180
1.2570g/mL
MFCD00000502
1M Solution in Dichloromethane
Chlorotrimethylsilane
100mL
AcroSeal™ Glass Bottle in Tin Can
Clear colorless solution
75-09-2,75-77-4
C3H9ClSi
(CH3)3SiCl

Chemical Identifiers

75-77-4,75-09-2
(CH3)3SiCl
MFCD00000502
C3H9ClSi
108.64
Chlorotrimethylsilane, 1M Solution in Dichloromethane

Safety and Handling

SDS & Safety Documents

GHS P Statement Suspected of causing cancer. Causes severe skin burns and eye damage. Harmful if swallowed. Flammable liquid and vapor. May cause drowsiness or dizziness. May cause damage to organs through prolonged o

GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EY

Danger