missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

chlorotrimethylsilane, 1M Solution in Dichloromethane, AcroSeal™, ACROS Organics™

$46.55 - $46.55

Chemical Identifiers

CAS 75-77-4,75-09-2
Molecular Formula C3H9ClSi
Linear Formula (CH3)3SiCl
Formula Weight 108.64
MDL Number MFCD00000502
Chemical Name or Material Chlorotrimethylsilane, 1M Solution in Dichloromethane
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC381601000 SDS Thermo Fisher Scientific
AC381601000
AcroSeal™ Glass Bottle in Tin Can 100mL Each for $46.55
Specifications

Description

Silylation reagent
Description & Specifications

Chemical Identifiers

75-77-4,75-09-2
(CH3)3SiCl
MFCD00000502
C3H9ClSi
108.64
Chlorotrimethylsilane, 1M Solution in Dichloromethane

Specifications

108.64
15, 2180
1.2570g/mL
MFCD00000502
C3H9ClSi
Chlorotrimethylsilane, 1M Solution in Dichloromethane
100mL
AcroSeal™ Glass Bottle in Tin Can
Clear colorless solution
75-09-2,75-77-4
(CH3)3SiCl