Learn More
Chlorotriethylsilane, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1554730
Description
Chlorotriethylsilane is used in laboratory chemicals. It is used for preparing silylating agent and acts as a catalyst for lewis acid. It is used for executing chemical reactions for silicon oil, silicon resin and polysiloxane. It is also used for eradicating robe agent, and used as protective agent and leather surface treating agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsChlorotriethylsilane is used in laboratory chemicals. It is used for preparing silylating agent and acts as a catalyst for lewis acid. It is used for executing chemical reactions for silicon oil, silicon resin and polysiloxane. It is also used for eradicating robe agent, and used as protective agent and leather surface treating agent.
Solubility
Soluble in aprotic organic solvents.
Notes
Incompatible with bases, strong oxidizing agents, strong acids, alcohols, amines, ammonia, aldehydes and acetone.
Specifications
145°C to 147°C | |
∼-75°C | |
250 g | |
(CH3CH2)3SiCl | |
UN2986 | |
29°C (84°F) | |
Soluble in aprotic organic solvents. | |
CC[Si](Cl)(CC)CC | |
150.72 | |
150.73 | |
Pungent | |
Moisture sensitive |
Chlorotriethylsilane | |
994-30-9 | |
C6H15ClSi | |
MFCD00000507 | |
1732860 | |
triethylchlorosilane, silane, chlorotriethyl, triethylsilyl chloride, chloro triethyl silane, unii-q1a687m1py, triethyl silyl chloride, triethychlorosilane, chlorotrietylsilane, clsiet3, et3sicl | |
DCFKHNIGBAHNSS-UHFFFAOYSA-N | |
chloro(triethyl)silane | |
13819 | |
≥98% | |
1.431 | |
0.899 |
Chemical Identifiers
994-30-9 | |
150.72 | |
DCFKHNIGBAHNSS-UHFFFAOYSA-N | |
13819 | |
CC[Si](Cl)(CC)CC |
C6H15ClSi | |
MFCD00000507 | |
triethylchlorosilane, silane, chlorotriethyl, triethylsilyl chloride, chloro triethyl silane, unii-q1a687m1py, triethyl silyl chloride, triethychlorosilane, chlorotrietylsilane, clsiet3, et3sicl | |
chloro(triethyl)silane |
Safety and Handling
GHS H Statement
H314-H318-H226-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Harmful if swallowed.
P210-P233-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H226-H312+H332-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, FLAMMABLE, N.O.S.
EINECSNumber : 213-615-6
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only