Please login to your online account to display your discounted pricing

Chlorotriethylgermane, 96%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Chlorotriethylgermane
Name Note 96%
CAS 994-28-5
Assay ≥95% (ex Cl)
Assay Percent Range 96%
Molecular Formula C6H15ClGe
Linear Formula (C2H5)3GeCl
Formula Weight 195.23
MDL Number MFCD00013591
Synonym Triethylgermanium chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC353190050

  SDS
thermo fisher scientific
353190050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC353190010

  SDS
thermo fisher scientific
353190010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥95% (ex Cl)
Formula Weight 195.23
Physical Form Liquid
Quantity 5g
Color Clear colorless
EINECS Number 213-614-0
Refractive Index 1.4580 to 1.4610 (20°C, 589nm)
Packaging Glass bottle
Name Note 96%
Molecular Formula C6H15ClGe
Linear Formula (C2H5)3GeCl
MDL Number MFCD00013591
Chemical Name or Material Chlorotriethylgermane
Assay Percent Range 96%
CAS 994-28-5
Synonym Triethylgermanium chloride
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
May cause respiratory irritation.
Causes skin irritation.
Harmful if inhaled.
Harmful if swallowed.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 GHS Signal Word: Warning
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
May cause respiratory irritation.
Causes skin irritation.
Harmful if inhaled.
Harmful if swallowed.
Causes serious eye irritation.