Please login to your online account to display your discounted pricing

Chlorotriethylgermane, 96%, ACROS Organics™


Quantity 1g

Manufacturer: thermo fisher scientific  353190010

 View more versions of this product

Catalog No. AC353190010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥95% (ex Cl)
Formula Weight 195.23
Quantity 1g
Color Clear colorless
EINECS Number 213-614-0
Chemical Name or Material Chlorotriethylgermane
Packaging Glass Bottle
Name Note 96%
Assay Percent Range 96%
Molecular Formula C6H15ClGe
MDL Number MFCD00013591
CAS 994-28-5
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
May cause respiratory irritation.
Causes skin irritation.
Harmful if inhaled.
Harmful if swallowed.
Causes serious eye irritation.
Refractive Index 1.4580 to 1.4610 (20°C, 589nm)
Linear Formula (C2H5)3GeCl
Synonym Triethylgermanium chloride
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic

Specifications

Chemical Name or Material Chlorotriethylgermane
Name Note 96%
CAS 994-28-5
Assay ≥95% (ex Cl)
Assay Percent Range 96%
Molecular Formula C6H15ClGe
Linear Formula (C2H5)3GeCl
Formula Weight 195.23
MDL Number MFCD00013591
Synonym Triethylgermanium chloride
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
May cause respiratory irritation.
Causes skin irritation.
Harmful if inhaled.
Harmful if swallowed.
Causes serious eye irritation.

SDS & Safety Documents