Please login to your online account to display your discounted pricing

Alfa Aesar™ Chlorotri-n-propylsilane, 98%

Chemical Identifiers

Chemical Name or Material Chlorotri-n-propylsilane
CAS 995-25-5
Molecular Formula (CH3CH2CH2)3SiCl
Formula Weight 192.81
MDL Number MFCD00000508
UN Number UN2987
Beilstein 1737489
Synonym Tri-n-propylchlorosilane; Tri-n-propylsilyl chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2206814

  SDS
alfa aesar™
B2206814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2206806

  SDS
alfa aesar™
B2206806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2206822

  SDS
alfa aesar™
B2206822
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 192.81
Synonym Tri-n-propylchlorosilane; Tri-n-propylsilyl chloride
Boiling Point 199° to 201°C
Quantity 25g
Odor Acrid
UN Number UN2987
TSCA TSCA
Health Hazard 2 GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, N.O.S.
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00000508
Molecular Formula (CH3CH2CH2)3SiCl
Health Hazard 1 Danger
Assay 98%
CAS 995-25-5
Chemical Name or Material Chlorotri-n-propylsilane
Beilstein 1737489
Flash Point 70°C (158°F)
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.